Assay Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232924.5 | XP_001232925.3 | 6 - 7 | 490 | 81 | |
| XM_025143032.1 | XP_024998800.1 | 6 - 7 | 503 | 81 | ||
| GenBank mRNA | BX930951.2 | - | 6 - 7 | 485 | 81 | |
| CR352489.1 | - | 6 - 7 | 520 | 81 | ||
More Information
Set Membership: |
|

TMEM50A
Genomic Map