Assay Details
| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934549.3 | XP_004934606.1 | 2 | 1686 | 101 | |
| XM_015301374.2 | XP_015156860.1 | 2 | 1689 | 101 | ||
| XM_025145283.1 | XP_025001051.1 | 2 | 2397 | 101 | ||
| XM_025145295.1 | XP_025001063.1 | 2 | 1694 | 101 | ||
| XM_025145308.1 | XP_025001076.1 | 2 | 1675 | 101 | ||
| XM_425554.6 | XP_425554.2 | 2 | 1742 | 101 | ||
More Information
Set Membership: |
|

KCNJ15
Genomic Map