Assay Details
| Gene Symbol: BCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275441.2 | XP_015130927.1 | 17 - 18 | 2015 | 74 | |
| XM_025155520.1 | XP_025011288.1 | 17 - 18 | 1833 | 74 | ||
| XM_025155521.1 | XP_025011289.1 | 17 - 18 | 2113 | 74 | ||
| XM_415244.5 | XP_415244.3 | 17 - 18 | 3689 | 74 | ||
More Information
Set Membership: |
|

BCR
Genomic Map