Assay Details
| Gene Symbol: BNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279006.2 | XP_015134492.1 | 12 | 5494 | 88 | |
| XM_413765.6 | XP_413765.1 | 12 | 5417 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BNIP2| Gene Symbol: BNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279006.2 | XP_015134492.1 | 12 | 5494 | 88 | |
| XM_413765.6 | XP_413765.1 | 12 | 5417 | 88 | ||
Set Membership: |
|