Assay Details
| Gene Symbol: LOC424588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291283.2 | XP_015146769.1 | 12 | 1650 | 80 | |
| XM_015291285.2 | XP_015146771.1 | 12 | 1656 | 80 | ||
| XM_025153082.1 | XP_025008850.1 | 12 | 1429 | 80 | ||
| XM_422428.6 | XP_422428.3 | 11 | 1606 | 80 | ||
More Information
Set Membership: |
|

LOC424588
Genomic Map