Assay Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285678.2 | XP_015141164.1 | 7 | 6476 | 89 | |
| XM_015285679.2 | XP_015141165.1 | 7 | 8832 | 89 | ||
| XM_015285680.2 | XP_015141166.1 | 6 | 6524 | 89 | ||
| XM_015285681.2 | XP_015141167.1 | 6 | 6506 | 89 | ||
| XM_025150037.1 | XP_025005805.1 | 8 | 6485 | 89 | ||
More Information
Set Membership: |
|

NWD2
Genomic Map