Assay Details
| Gene Symbol: SMIM10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945308.3 | XP_004945365.1 | 5 | 14589 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM10L1| Gene Symbol: SMIM10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945308.3 | XP_004945365.1 | 5 | 14589 | 79 | |
Set Membership: |
|