Assay Details
| Gene Symbol: VEGFRKDRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278582.2 | XP_015134068.1 | 30 | 7917 | 115 | |
| XM_015278583.2 | XP_015134069.1 | 30 | 7915 | 115 | ||
| XM_025149942.1 | XP_025005710.1 | 29 | 7808 | 115 | ||
| XM_420292.6 | XP_420292.4 | 30 | 7913 | 115 | ||
More Information
Set Membership: |
|

VEGFRKDRL
Genomic Map