Assay Details
| Gene Symbol: C23H1ORF109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417759.6 | XP_417759.5 | 3 - 4 | 575 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C23H1ORF109| Gene Symbol: C23H1ORF109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417759.6 | XP_417759.5 | 3 - 4 | 575 | 145 | |
Set Membership: |
|