Assay Details
| Gene Symbol: TMEM165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285536.2 | XP_015141022.1 | 3 - 4 | 1277 | 64 | |
| XM_015285537.2 | XP_015141023.1 | 4 - 5 | 904 | 64 | ||
More Information
Set Membership: |
|

TMEM165
Genomic Map