Assay Details
| Gene Symbol: CXorf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640516.4 | XP_003640564.2 | 4 - 5 | 1602 | 109 | |
| XM_015272723.2 | XP_015128209.1 | 4 - 5 | 1490 | 109 | ||
More Information
Set Membership: |
|

CXorf23
Genomic Map