Assay Details
| Gene Symbol: CXorf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640516.4 | XP_003640564.2 | 6 - 7 | 1899 | 137 | |
| XM_015272723.2 | XP_015128209.1 | 6 - 7 | 1787 | 137 | ||
| XM_025146578.1 | XP_025002346.1 | 6 - 7 | 1728 | 137 | ||
More Information
Set Membership: |
|

CXorf23
Genomic Map