Assay Details
| Gene Symbol: NCKAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289932.2 | XP_015145418.1 | 14 - 15 | 5401 | 109 | |
| XM_015289933.2 | XP_015145419.1 | 14 - 15 | 5299 | 109 | ||
| XM_015289934.1 | XP_015145420.1 | 10 - 11 | 4919 | 109 | ||
| XM_015289935.1 | XP_015145421.1 | 13 - 14 | 5226 | 109 | ||
| XM_025152641.1 | XP_025008409.1 | 14 - 15 | 5402 | 109 | ||
| XM_025152642.1 | XP_025008410.1 | 14 - 15 | 5343 | 109 | ||
| XM_025152643.1 | XP_025008411.1 | 14 - 15 | 5402 | 109 | ||
More Information
Set Membership: |
|

NCKAP5
Genomic Map