Assay Details
| Gene Symbol: LOC101751545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290807.2 | XP_015146293.1 | 4 - 5 | 853 | 104 | |
| XM_015290812.2 | XP_015146298.1 | 3 - 4 | 667 | 104 | ||
| XM_015290826.2 | XP_015146312.1 | 2 - 3 | 569 | 104 | ||
| XM_015290832.2 | XP_015146318.1 | 2 - 3 | 539 | 104 | ||
| XM_015290837.2 | XP_015146323.1 | 3 - 4 | 707 | 104 | ||
| GenBank mRNA | KC818437.1 | - | 1 - 2 | 405 | 104 | |
More Information
Set Membership: |
|

LOC101751545
Genomic Map