Assay Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233247.4 | XP_001233248.2 | 8 - 9 | 4633 | 62 | |
| XM_004949268.3 | XP_004949325.1 | 7 - 8 | 4438 | 62 | ||
More Information
Set Membership: |
|

SLC27A6
Genomic Map