Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300287.2 | XP_015155773.1 | 7 - 8 | 860 | 58 | |
| XM_015300288.2 | XP_015155774.1 | 7 - 8 | 859 | 58 | ||
| XM_015300289.2 | XP_015155775.1 | 7 - 8 | 858 | 58 | ||
More Information
Set Membership: |
|

CS
Genomic Map