Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294380.2 | XP_015149866.1 | 15 - 16 | 6438 | 103 | |
| XM_015294382.2 | XP_015149868.2 | 11 - 12 | 1304 | 103 | ||
| XM_015294383.2 | XP_015149869.1 | 16 - 17 | 6483 | 103 | ||
| XM_025155364.1 | XP_025011132.1 | 16 - 17 | 7509 | 103 | ||
| XM_025155365.1 | XP_025011133.1 | 13 - 14 | 1586 | 103 | ||
| XM_025155366.1 | XP_025011134.1 | 14 - 15 | 6294 | 103 | ||
| XM_414805.6 | XP_414805.3 | 12 - 13 | 1418 | 103 | ||
More Information
Set Membership: |
|

USP51
Genomic Map