Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294380.2 | XP_015149866.1 | 17 - 18 | 6651 | 94 | |
| XM_015294382.2 | XP_015149868.2 | 13 - 14 | 1517 | 94 | ||
| XM_015294383.2 | XP_015149869.1 | 18 - 19 | 6696 | 94 | ||
| XM_025155364.1 | XP_025011132.1 | 18 - 19 | 7722 | 94 | ||
| XM_025155365.1 | XP_025011133.1 | 15 - 16 | 1799 | 94 | ||
| XM_025155366.1 | XP_025011134.1 | 16 - 17 | 6507 | 94 | ||
| XM_414805.6 | XP_414805.3 | 14 - 15 | 1631 | 94 | ||
More Information
Set Membership: |
|

USP51
Genomic Map