Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294382.2 | XP_015149868.2 | 3 - 4 | 358 | 101 | |
| XM_015294383.2 | XP_015149869.1 | 7 - 8 | 5393 | 101 | ||
| XM_025155364.1 | XP_025011132.1 | 7 - 8 | 6419 | 101 | ||
| XM_025155365.1 | XP_025011133.1 | 4 - 5 | 496 | 101 | ||
| XM_414805.6 | XP_414805.3 | 3 - 4 | 328 | 101 | ||
More Information
Set Membership: |
|

USP51
Genomic Map