Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294382.2 | XP_015149868.2 | 4 - 5 | 400 | 77 | |
| XM_015294383.2 | XP_015149869.1 | 8 - 9 | 5435 | 77 | ||
| XM_025155364.1 | XP_025011132.1 | 8 - 9 | 6461 | 77 | ||
| XM_025155365.1 | XP_025011133.1 | 5 - 6 | 538 | 77 | ||
| XM_414805.6 | XP_414805.3 | 4 - 5 | 370 | 77 | ||
More Information
Set Membership: |
|

USP51
Genomic Map