Assay Details
| Gene Symbol: TUBA8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416397.4 | XP_416397.1 | 1 - 2 | 875 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_416397.4 transcripts.
Set Membership: |
|

TUBA8A
Genomic Map