Assay Details
| Gene Symbol: GLTSCR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935345.3 | XP_004935402.1 | 12 | 16120 | 105 | |
| XM_015283843.2 | XP_015139329.1 | 12 | 4481 | 105 | ||
| XM_015283844.2 | XP_015139330.1 | 13 | 16038 | 105 | ||
| XM_015283845.2 | XP_015139331.1 | 13 | 4609 | 105 | ||
| XM_015283846.2 | XP_015139332.1 | 9 | 4026 | 105 | ||
| XM_025148871.1 | XP_025004639.1 | 13 | 14491 | 105 | ||
| XM_025148872.1 | XP_025004640.1 | 12 | 6238 | 105 | ||
More Information
Set Membership: |
|

GLTSCR1L
Genomic Map