Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281788.2 | XP_015137274.1 | 4 - 5 | 678 | 88 | |
| XM_025148019.1 | XP_025003787.1 | 4 - 5 | 658 | 88 | ||
| XM_025148020.1 | XP_025003788.1 | 4 - 5 | 658 | 88 | ||
| XM_418759.5 | XP_418759.4 | 4 - 5 | 658 | 88 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map