Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155120.1 | XP_025010888.1 | 4 - 5 | 449 | 70 | |
| XM_025155125.1 | XP_025010893.1 | 4 - 5 | 445 | 70 | ||
More Information
Set Membership: |
|

PMM1
Genomic Map