Assay Details
| Gene Symbol: GCOG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154246.1 | XP_025010014.1 | 3 - 4 | 1382 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCOG8| Gene Symbol: GCOG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154246.1 | XP_025010014.1 | 3 - 4 | 1382 | 76 | |
Set Membership: |
|