Assay Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276773.2 | XP_015132259.1 | 7 | 3434 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR137C| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276773.2 | XP_015132259.1 | 7 | 3434 | 117 | |
Set Membership: |
|