Assay Details
| Gene Symbol: PTPRVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298717.2 | XP_015154203.2 | 10448 | 106 | ||
| XM_015298719.1 | XP_015154205.1 | 5338 | 106 | |||
| XM_025143848.1 | XP_024999616.1 | 10175 | 106 | |||
More Information
Set Membership: |
|

PTPRVP
Genomic Map