Assay Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154265.1 | XP_025010033.1 | 4 - 5 | 1085 | 103 | |
XM_025154266.1 | XP_025010034.1 | 3 - 4 | 944 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
