Assay Details
| Gene Symbol: TOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296795.2 | XP_015152281.1 | 8 | 2030 | 95 | |
| XM_015296796.2 | XP_015152282.1 | 9 | 2883 | 95 | ||
| XM_025142323.1 | XP_024998091.1 | 9 | 2049 | 95 | ||
| XM_417340.6 | XP_417340.3 | 9 | 2096 | 95 | ||
More Information
Set Membership: |
|

TOX2
Genomic Map