Assay Details
| Gene Symbol: LOC430433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015272637.2 | XP_015128123.1 | 2 - 3 | 525 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015272637.2 transcripts.
Set Membership: |
|

LOC430433
Genomic Map