Assay Details
| Gene Symbol: PPM1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640351.4 | XP_003640399.3 | 10 | 6893 | 111 | |
| XM_004937558.3 | XP_004937615.1 | 11 | 3505 | 111 | ||
| XM_015282711.2 | XP_015138197.1 | 10 | 3583 | 111 | ||
More Information
Set Membership: |
|

PPM1H
Genomic Map