Assay Details
| Gene Symbol: TFDP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938530.3 | XP_004938587.1 | 9 - 10 | 1098 | 97 | |
| XM_015277902.2 | XP_015133388.1 | 10 - 11 | 1238 | 97 | ||
| XM_015277903.2 | XP_015133389.1 | 10 - 11 | 1240 | 97 | ||
| XM_015277904.2 | XP_015133390.1 | 10 - 11 | 1235 | 97 | ||
| XM_015277905.2 | XP_015133391.1 | 10 - 11 | 1237 | 97 | ||
More Information
Set Membership: |
|

TFDP3
Genomic Map