Assay Details
| Gene Symbol: FAM188B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418488.6 | XP_418488.5 | 15 - 16 | 2063 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM188B| Gene Symbol: FAM188B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418488.6 | XP_418488.5 | 15 - 16 | 2063 | 60 | |
Set Membership: |
|