Assay Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940609.2 | XP_004940666.1 | 5 - 6 | 1338 | 69 | |
| XM_426256.6 | XP_426256.3 | 5 - 6 | 922 | 69 | ||
More Information
Set Membership: |
|

RBM41
Genomic Map