Assay Details
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275576.2 | XP_015131062.2 | 2 - 3 | 1597 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015275576.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
