Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292254.2 | XP_015147740.1 | 3 - 4 | 586 | 96 | |
| XM_015292255.2 | XP_015147741.1 | 3 - 4 | 547 | 96 | ||
More Information
Set Membership: |
|

PARP16
Genomic Map