Assay Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945619.3 | XP_004945676.1 | 4 - 5 | 603 | 76 | |
| XM_415191.6 | XP_415191.2 | 4 - 5 | 548 | 76 | ||
More Information
Set Membership: |
|

KCTD10
Genomic Map