Assay Details
| Gene Symbol: FAM228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285041.2 | XP_015140527.1 | 4 - 5 | 440 | 99 | |
| XM_015285042.2 | XP_015140528.1 | 4 - 5 | 387 | 99 | ||
| XM_025149106.1 | XP_025004874.1 | 5 - 6 | 733 | 99 | ||
More Information
Set Membership: |
|

FAM228
Genomic Map