Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299088.2 | XP_015154574.1 | 1149 | 78 | ||
| XM_015299089.2 | XP_015154575.1 | 1154 | 78 | |||
| XM_015299090.2 | XP_015154576.1 | 960 | 78 | |||
| XM_025143901.1 | XP_024999669.1 | 1683 | 78 | |||
| XM_417961.5 | XP_417961.2 | 1746 | 78 | |||
More Information
Set Membership: |
|

TMCC2
Genomic Map