Assay Details
| Gene Symbol: GPR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425941.6 | XP_425941.2 | 1 - 2 | 169 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_425941.6 transcripts.
Set Membership: |
|

GPR20
Genomic Map