Assay Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284934.2 | XP_015140420.1 | 4 - 5 | 665 | 61 | |
| XM_015284935.2 | XP_015140421.1 | 4 - 5 | 7824 | 61 | ||
| XM_025148866.1 | XP_025004634.1 | 4 - 5 | 7824 | 61 | ||
More Information
Set Membership: |
|

FAM150B
Genomic Map