Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289876.2 | XP_015145362.1 | 14 | 8425 | 98 | |
| XM_015289877.2 | XP_015145363.1 | 16 | 8188 | 98 | ||
| XM_015289878.2 | XP_015145364.1 | 16 | 8058 | 98 | ||
| XM_015289881.2 | XP_015145367.1 | 16 | 8353 | 98 | ||
| XM_015289882.2 | XP_015145368.1 | 16 | 8043 | 98 | ||
| XM_015289883.2 | XP_015145369.1 | 14 | 8407 | 98 | ||
| XM_015289884.2 | XP_015145370.1 | 14 | 8368 | 98 | ||
| XM_015289885.2 | XP_015145371.1 | 15 | 7781 | 98 | ||
| XM_025152449.1 | XP_025008217.1 | 15 | 8483 | 98 | ||
| XM_025152451.1 | XP_025008219.1 | 15 | 8000 | 98 | ||
| XM_025152452.1 | XP_025008220.1 | 15 | 7985 | 98 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map