Assay Details
| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414402.6 | XP_414402.2 | 1 - 2 | 120 | 92 | |
| GenBank mRNA | BX931060.1 | - | 1 - 2 | 97 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPP14| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414402.6 | XP_414402.2 | 1 - 2 | 120 | 92 | |
| GenBank mRNA | BX931060.1 | - | 1 - 2 | 97 | 92 | |
Set Membership: |
|