Assay Details
| Gene Symbol: RPP25L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424977.5 | XP_424977.1 | 1 - 2 | 39 | 50 | |
| GenBank mRNA | BX931336.1 | - | 1 - 2 | 23 | 50 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_424977.5 transcripts.
Set Membership: |
|

RPP25L
Genomic Map