Assay Details
| Gene Symbol: PDLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942091.3 | XP_004942148.1 | 5 - 6 | 3757 | 94 | |
| XM_025151869.1 | XP_025007637.1 | 3 - 4 | 699 | 94 | ||
| XM_426503.6 | XP_426503.4 | 5 - 6 | 738 | 94 | ||
| GenBank mRNA | BX931480.2 | - | 5 - 6 | 744 | 94 | |
| BX932965.1 | - | 634 | 94 | |||
| CR387509.1 | - | 5 - 6 | 711 | 94 | ||
More Information
Set Membership: |
|

PDLIM1
Genomic Map