Assay Details
| Gene Symbol: MED28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641248.4 | XP_003641296.3 | 1 - 2 | 173 | 76 | |
| GenBank mRNA | BX931501.1 | - | 1 - 2 | 175 | 76 | |
More Information
Set Membership: |
|

MED28
Genomic Map