Assay Details
| Gene Symbol: COX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276400.2 | XP_015131886.1 | 4 - 5 | 696 | 94 | |
| XM_015276401.2 | XP_015131887.1 | 4 - 5 | 700 | 94 | ||
| GenBank mRNA | BX931540.1 | - | 3 - 4 | 842 | 94 | |
More Information
Set Membership: |
|

COX18
Genomic Map