Assay Details
| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424417.6 | XP_424417.3 | 2 - 3 | 220 | 73 | |
| GenBank mRNA | BX931577.2 | - | 1 - 2 | 264 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRHPR| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424417.6 | XP_424417.3 | 2 - 3 | 220 | 73 | |
| GenBank mRNA | BX931577.2 | - | 1 - 2 | 264 | 73 | |
Set Membership: |
|