Assay Details
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025142950.1 | XP_024998718.1 | 1 - 2 | 157 | 70 | |
XM_424381.6 | XP_424381.4 | 1 - 2 | 120 | 70 | ||
GenBank mRNA | BX931594.2 | - | 1 - 2 | 108 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
