Assay Details
| Gene Symbol: USB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292315.2 | XP_015147801.1 | 4 - 5 | 524 | 74 | |
| XM_015292317.2 | XP_015147803.1 | 3 - 4 | 374 | 74 | ||
| XM_015292318.2 | XP_015147804.1 | 4 - 5 | 491 | 74 | ||
| XM_015292319.2 | XP_015147805.1 | 4 - 5 | 491 | 74 | ||
| XM_025154275.1 | XP_025010043.1 | 3 - 4 | 598 | 74 | ||
| XM_025154276.1 | XP_025010044.1 | 3 - 4 | 600 | 74 | ||
| XM_025154277.1 | XP_025010045.1 | 3 - 4 | 591 | 74 | ||
| XM_025154278.1 | XP_025010046.1 | 3 - 4 | 596 | 74 | ||
More Information
Set Membership: |
|

USB1
Genomic Map